Reagents Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C1324500-10KG| Item: 6228308

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white crystals
  • Density: 1.6
  • Melting point: 75 °C (167 °F), decomposes
  • Solubility information: 144 g/L at 25 °C (77 °F)
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNa3PO4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium phosphate tribasic 12-hydrate, TSP, Trisodium phosphate dodecahydrate
  • FormSolid
  • Weight (g)10000
  • Formula Weight163.94
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10101-89-0
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionSodium Phosphate Tribasic Dodecahydrate, ACS Reagent, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A