Reagents Sodium Phosphate Monobasic Monohydrate, ACS Reagent, Natural Poly Container, 3 kg

Representative image only.

Reagents Sodium Phosphate Monobasic Monohydrate, ACS Reagent, Natural Poly Container, 3 kg

Reagents | Mfr: C1323500-3A5| Item: 6228306

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white crystals
  • Density: 2.04
  • Boiling point: approximately 204 °C (399.2 °F)
  • Solubility information: 949 g/L at 25 °C (77 °F)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaH2PO4·H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium phosphate monobasic 1-hydrate, Sodium dihydrogen phosphate monohydrate
  • FormSolid
  • Weight (g)3000
  • Formula Weight137.99
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10049-21-5
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)100
  • Melting Point F (° F)212
  • DescriptionSodium Phosphate Monobasic Monohydrate, ACS Reagent, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A