Reagents Sodium Phosphate Dibasic Anhydrous, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Phosphate Dibasic Anhydrous, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2324000-10KG| Item: 6228592

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white granules
  • Density: 2.07
  • Melting point: >240 °C (464 °F), decomposes
  • Solubility information: 118 g/L at 25 °C (77 °F)
  • pH range: 8.7 to 9.3
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2HPO4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsDisodium hydrogen phosphate
  • FormSolid
  • Weight (g)10000
  • Formula Weight141.96
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7558-79-4
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Phosphate Dibasic Anhydrous, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A