Reagents Sodium Phosphate Dibasic Anhydrous, ACS Reagent, Poly Pail, 12 kg

Representative image only.

Reagents Sodium Phosphate Dibasic Anhydrous, ACS Reagent, Poly Pail, 12 kg

Reagents | Mfr: C1324050-12A5| Item: 6228307

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 2.07
  • Melting poing: >240 °C (464 °F), decomposes
  • Solubility information: 118 g/L at 25 °C (77 °F)
  • pH range: 8.7 to 9.3
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNa2HPO4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsDisodium hydrogen phosphate
  • FormSolid
  • Weight (g)12000
  • PackagingPoly pail
  • CAS Number7558-79-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Phosphate Dibasic Anhydrous, ACS Reagent, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A