Reagents Sodium Persulfate, Reagent, Natural Poly Container, 2.25 kg

Representative image only.

Reagents Sodium Persulfate, Reagent, Natural Poly Container, 2.25 kg

Reagents | Mfr: C2323100-2.25A5| Item: 6228589

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.6
  • Solubility information: 556 g/L at 20 °C (68 °F)
  • 2.25 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeReagent
  • Linear FormulaNa2S2O8
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsSodium peroxydisulfate
  • Weight (g)2250
  • Formula Weight238.1
  • PackagingPoly container
  • DOT InformationSODIUM PERSULFATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number7775-27-1
  • UN NumberUN1505
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)120
  • Melting Point F (° F)248
  • DescriptionSodium Persulfate, Reagent, Natural Poly Container, 2.25 kg
Documents & Certificates
Reviews
Q&A