Reagents Sodium Nitrate, ACS Reagent, Fiber Drum, 10 kg

Representative image only.

Reagents Sodium Nitrate, ACS Reagent, Fiber Drum, 10 kg

Reagents | Mfr: C1320500-10F4| Item: 6228304

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 2.26
  • Boiling point: 380 °C (716 °F) with decomposition
  • Solubility information: 874 g/L at 20 °C (68 °F)
  • pH range: 5.5 to 8.3
  • 10 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • ClassSalts
  • ChemistryInorganics
  • Weight (g)10000
  • PackagingFiber drum
  • Melting Point C (° C)306
  • Melting Point F (° F)582.8
  • DescriptionSodium Nitrate, ACS Reagent, Fiber Drum, 10 kg
Documents & Certificates
Reviews
Q&A