Reagents Sodium Metabisulfite, Purified, Fiber Drum, 50 lb

Representative image only.

Reagents Sodium Metabisulfite, Purified, Fiber Drum, 50 lb

Reagents | Mfr: C2320100-50F5| Item: 6228585

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.48
  • Melting point: 150 °C (302 °F), decomposes
  • Solubility information: 667 g/L at 25 °C (77 °F)
  • 50 lb fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2O5S2
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)22680
  • Formula Weight190.11
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7681-57-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Metabisulfite, Purified, Fiber Drum, 50 lb
Documents & Certificates
Reviews
Q&A