Reagents Sodium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125g

Representative image only.

Reagents Sodium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125g

Reagents | Mfr: C1320000-125D2| Item: 6228303

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • White granules
  • Density: 3.67
  • Solubility information: 1840 g/L at 25 °C (77 °F)
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaI
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • Formula Weight149.89
  • Vapor Pressure (torr)1
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7681-82-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)1300
  • Boiling Point F (° F)2372
  • Melting Point C (° C)661
  • Melting Point F (° F)1221.8
  • DescriptionSodium Iodide, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125g
Documents & Certificates
Reviews
Q&A