Reagents Sodium Hydroxide, ACS Reagent, Natural Poly Container, 1 kg

Representative image only.

Reagents Sodium Hydroxide, ACS Reagent, Natural Poly Container, 1 kg

Reagents | Mfr: C1318000-1KG| Item: 6228298

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 97%
  • White pellets
  • Density: 2.13
  • Solubility information: 1000 g/L at 25 °C (77 °F)
  • 1 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaOH
  • ClassBase
  • ChemistryInorganics
  • SynonymsCaustic soda, Lye
  • FormSolid
  • Weight (g)1000
  • Formula Weight40
  • PackagingPoly container
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)1390
  • Boiling Point F (° F)2534
  • Melting Point C (° C)318
  • Melting Point F (° F)604.4
  • DescriptionSodium Hydroxide, ACS Reagent, Natural Poly Container, 1 kg
Documents & Certificates
Reviews
Q&A