Reagents Sodium Hydroxide, 50% (w/w), Reagent, Poly Drum, 200 L

Representative image only.

Reagents Sodium Hydroxide, 50% (w/w), Reagent, Poly Drum, 200 L

Reagents | Mfr: C1318600-200E| Item: 6228302

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 1.53
  • Solubility information: miscible
  • pH range: 14
  • 200 L poly drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeReagent
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)200000
  • Vapor Pressure (torr)13
  • PackagingPoly drum
  • DOT InformationSODIUM HYDROXIDE SOLUTION, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number1310-73-2
  • UN NumberUN1824
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)140
  • Boiling Point F (° F)284
  • Melting Point C (° C)12
  • Melting Point F (° F)53.6
  • DescriptionSodium Hydroxide, 50% (w/w), Reagent, Poly Drum, 200 L
Documents & Certificates
Reviews
Q&A