Reagents Sodium Dodecyl Sulfate, Wide-Mouth Container, 100 g

Representative image only.

Reagents Sodium Dodecyl Sulfate, Wide-Mouth Container, 100 g

Reagents | Mfr: C2197800-100A6| Item: 6228499

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White powderDensity: 1.6
  • Melting point: 204 to 207 °C (399.2 to 404.6 °F)
  • Flash point: 150 °C (302 °F)
  • Solubility information: 150 g/L at 20 °C (68 °F)
  • 100 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaCH3(CH2)11OSO3Na
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsSodium lauryl sulfate, Dodecyl sodium sulfate, SDS
  • Weight (g)100
  • Formula Weight288.38
  • PackagingWide-mouth container
  • DOT InformationFLAMMABLE SOLIDS, ORGANIC, N.O.S. (SODIUM LAURYL SULFATE), 4.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number151-21-3
  • UN NumberUN1325
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Dodecyl Sulfate, Wide-Mouth Container, 100 g
Documents & Certificates
Reviews
Q&A