Reagents Sodium Dithionite, Purified, Natural Poly Container, 2.25 kg

Representative image only.

Reagents Sodium Dithionite, Purified, Natural Poly Container, 2.25 kg

Reagents | Mfr: C2316300-2.5A5| Item: 6228575

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White to off-white powder
  • Density: 0.88
  • Boiling point: >80 °C (176 °F), decomposes
  • Melting point: 52 °C (125.6 °F), decomposes
  • Flash point: >100 °C (>212 °F)
  • Solubility information: 241 g/L at 20 °C (68 °F)
  • 2.25 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2O4S2
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium hydrosulfite
  • FormSolid
  • Weight (g)2250
  • Formula Weight174.11
  • PackagingPoly container
  • DOT InformationSODIUM DITHIONITE, 4.2, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1384
  • CAS Number7775-14-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Dithionite, Purified, Natural Poly Container, 2.25 kg
Documents & Certificates
Reviews
Q&A