Reagents Sodium Chloride, Purified, Fiber Drum, 50 lb

Representative image only.

Reagents Sodium Chloride, Purified, Fiber Drum, 50 lb

Reagents | Mfr: C2313000-50F5| Item: 6228568

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white crystals
  • Density: 2.17
  • Solubility information: 360 g/L at 25 °C (68 °F)
  • pH range: 5 to 9
  • 50 lb fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNaCl
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)22680
  • Formula Weight58.44
  • Vapor Pressure (torr)1
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7647-14-5
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)1461
  • Boiling Point F (° F)2661.8
  • Melting Point C (° C)801
  • Melting Point F (° F)1473.8
  • DescriptionSodium Chloride, Purified, Fiber Drum, 50 lb
Documents & Certificates
Reviews
Q&A