Reagents Sodium Carbonate Anhydrous, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Carbonate Anhydrous, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2312100-10KG| Item: 6228566

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white granules
  • Density: 2.53
  • Boiling point: decomposes
  • Solubility Information: 215 g/L at 20 °C (68 °F)
  • pH range: 11 to 12
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2CO3
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSoda ash
  • FormSolid
  • Weight (g)10000
  • Formula Weight105.99
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number497-19-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)851
  • Melting Point F (° F)1563.8
  • DescriptionSodium Carbonate Anhydrous, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A