Reagents Sodium Carbonate Anhydrous, Purified, Fiber Drum, 100 lb

Representative image only.

Reagents Sodium Carbonate Anhydrous, Purified, Fiber Drum, 100 lb

Reagents | Mfr: C2312100-100LB| Item: 6228565

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white granules
  • Density: 2.53
  • Boiling point: decomposes
  • Solubility Information: 215 g/L at 20 °C (68 °F)
  • pH range: 11 to 12
  • 100 lb fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2CO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsSoda ash
  • Weight (g)45359
  • Formula Weight105.99
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number497-19-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)851
  • Melting Point F (° F)1563.8
  • DescriptionSodium Carbonate Anhydrous, Purified, Fiber Drum, 100 lb
Documents & Certificates
Reviews
Q&A