Reagents Sodium Bromide, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Sodium Bromide, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2311000-500A5| Item: 6228564

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white granules
  • Density: 3.21
  • Solubility information: 900 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNaBr
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)500
  • Formula Weight102.89
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7647-15-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)1390
  • Boiling Point F (° F)2534
  • Melting Point C (° C)755
  • Melting Point F (° F)1391
  • DescriptionSodium Bromide, Purified, Wide-Mouth Container, 500 g
Documents & Certificates
Reviews
Q&A