Reagents Sodium Borate, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Sodium Borate, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C1310500-10KG| Item: 6228282

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • White granules
  • Density: 1.73
  • Boiling point: approximately 320 °C (608 °F)
  • Solubility information: 31.7 g/L at 25 °C (77 °F)
  • pH range: 9.15 to 9.2
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNa2B4O7·10H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium borate decahydrate, Borax, Sodium tetraborate decahydrate
  • FormSolid
  • Weight (g)10000
  • Formula Weight381.37
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Melting Point C (° C)75
  • Melting Point F (° F)167
Documents & Certificates
Reviews
Q&A