Reagents Sodium Bisulfate Anhydrous, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Bisulfate Anhydrous, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C2309500-100C2| Item: 6228561

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.74
  • Solubility information: 285 g/L at 25 °C (77 °F)
  • 100 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaNaHSO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)100
  • Formula Weight120.06
  • PackagingWide-mouth amber glass bottle
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (SODIUM BISULFATE), 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3260
  • CAS Number7681-38-1
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)315
  • Melting Point F (° F)599
  • DescriptionSodium Bisulfate Anhydrous, Wide-Mouth Amber Glass Bottle, 100 g
Documents & Certificates
Reviews
Q&A