Reagents Silver Sulfate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents Silver Sulfate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C1304000-25C1| Item: 6228276

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum 98%
  • Colorless/white solid
  • Density: 5.45
  • Boiling point: 1085 °C (1985 °F), decomposes
  • Solubility information: 8.4 g/L at 25 °C (77 °F)
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaAg2SO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)25
  • Melting Point C (° C)652
  • Melting Point F (° F)1205.6
Documents & Certificates
Reviews
Q&A