Reagents Propanol (Normal), ACS Reagent, Amber Glass Bottle, 1 L

Representative image only.

Reagents Propanol (Normal), ACS Reagent, Amber Glass Bottle, 1 L

Reagents | Mfr: C1121200-1C| Item: 6228133

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 0.8
  • Flash point: 23 °C (73.4 °F)
  • Refractive index: 1.384 to 1.386nD
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCH3(CH2)2OH
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsN-Propyl Alcohol, n-propanol, 1-Propanol
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight60.1
  • Vapor Pressure (torr)14.251
  • PackagingAmber glass bottle
  • DOT InformationN-PROPANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number71-23-8
  • UN NumberUN1274
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)97
  • Boiling Point F (° F)206.6
  • Melting Point C (° C)-127
  • Melting Point F (° F)-196.6
  • DescriptionPropanol (Normal), ACS Reagent, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A