Reagents Potassium Thiocyanate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Representative image only.

Reagents Potassium Thiocyanate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Reagents | Mfr: C1293000-125D2| Item: 6228273

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 1.89
  • Boiling point: 500 °C (932 °F), decomposes
  • Melting point: 170 to 179 °C (338 to 354.2 °F)
  • Solubility information: 2380 g/L at 25 °C (77 °F)
  • pH range: 5.3 to 8.7
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKSCN
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • Formula Weight97.18
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number333-20-0
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPotassium Thiocyanate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g
Documents & Certificates
Reviews
Q&A