Reagents Potassium Sulfate, Low Nitrogen, Purified, Poly Pail, 12 kg

Representative image only.

Reagents Potassium Sulfate, Low Nitrogen, Purified, Poly Pail, 12 kg

Reagents | Mfr: C2291000-12KG| Item: 6228553

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.66
  • Solubility information: soluble
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaK2SO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)12000
  • Formula Weight174.26
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-80-5
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)1069
  • Melting Point F (° F)1956.2
  • DescriptionPotassium Sulfate, Low Nitrogen, Purified, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A