Reagents Potassium Sulfate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Representative image only.

Reagents Potassium Sulfate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Reagents | Mfr: C1291000-125D2| Item: 6228272

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 2.66
  • Solubility information: soluble
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaK2SO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • Formula Weight174.26
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-80-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)1689
  • Boiling Point F (° F)3072.2
  • Melting Point C (° C)1069
  • Melting Point F (° F)1956.2
  • DescriptionPotassium Sulfate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g
Documents & Certificates
Reviews
Q&A