Reagents Potassium Phosphate Monobasic, ACS Reagent, Poly Pail, 10 kg

Representative image only.

Reagents Potassium Phosphate Monobasic, ACS Reagent, Poly Pail, 10 kg

Reagents | Mfr: C1289000-10KG| Item: 6228265

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 2.34
  • Boiling point: 400 °C (752 °F), loses water and becomes metaphosphate
  • Solubility information: 250 g/L at 25 °C (77 °F)
  • pH range: 4.1 to 4.5
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKH2PO4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsPotassium dihydrogen phosphate
  • FormSolid
  • Weight (g)10000
  • Formula Weight136.09
  • Vapor Pressure (torr)7.3
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-77-0
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)252.6
  • Melting Point F (° F)486.68
  • DescriptionPotassium Phosphate Monobasic, ACS Reagent, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A