Reagents Potassium Permanganate, Purified, Poly Pail, 12 kg

Representative image only.

Reagents Potassium Permanganate, Purified, Poly Pail, 12 kg

Reagents | Mfr: C2288500-12KG| Item: 6228550

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Purple-black solid
  • Density: 2.7
  • Melting point: >240 °C (>464 °F)
  • Solubility information: 76 g/L at 25 °C (77 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaKMnO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)12000
  • Formula Weight158.03
  • PackagingPoly pail
  • DOT InformationPOTASSIUM PERMANGANATE, 5.1, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1490
  • CAS Number7722-64-7
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Permanganate, Purified, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A