Reagents Potassium Nitrate, Amber Glass Bottle, 250 mL

Representative image only.

Reagents Potassium Nitrate, Amber Glass Bottle, 250 mL

Reagents | Mfr: C7286500-250C| Item: 6228806

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Saturated, Aqueous
  • Colorless liquid
  • Density: 1.2
  • Boiling point: approximately 100 °C (212 °F)
  • Solubility information: miscible
  • pH range: 4.5 to 8.5
  • 250 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Temperature changes may cause precipitation.

Specifications
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)250
  • PackagingAmber glass bottle
  • DOT InformationNITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S. (POTASSIUM NITRATE), 5.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3218
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Melting Point C (° C)334
  • Melting Point F (° F)633.2
  • DescriptionPotassium Nitrate, Amber Glass Bottle, 250 mL
Documents & Certificates
Reviews
Q&A