Reagents Potassium Iodide, 1.0N, Volumetric, Amber Black Poly Bottle, 4 L

Representative image only.

Reagents Potassium Iodide, 1.0N, Volumetric, Amber Black Poly Bottle, 4 L

Reagents | Mfr: C7286000-4B| Item: 6228803

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 16.6% (w/v), (Unstandardized)
  • Colorless to slightly yellow liquid
  • Density: 1.12
  • Boiling point: approximately 100 °C (212 °F)
  • Solubility information: miscible
  • pH range: 6 to 9.2
  • 4 L amber black poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Normality1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber black poly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)395
  • Shelf Life (months)12
  • DescriptionPotassium Iodide, 1.0N, Volumetric, Amber Black Poly Bottle, 4 L
Documents & Certificates
Reviews
Q&A