Reagents Potassium Hydroxide, Purified, Natural Poly Container, 3 kg

Representative image only.

Reagents Potassium Hydroxide, Purified, Natural Poly Container, 3 kg

Reagents | Mfr: C2285000-3A5| Item: 6228547

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White deliquescent solid
  • Density: 2.04
  • Boiling point: approximately 1320 °C (2408 °F)
  • Solubility information: 1210 g/L at 25 °C (77 °F)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaKOH
  • ClassBase
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • Formula Weight56.11
  • PackagingPoly container
  • DOT InformationPOTASSIUM HYDROXIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1813
  • CAS Number1310-58-3
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Hydroxide, Purified, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A