Reagents Potassium Hydroxide, 0.100N, Standardized, Cubitainer® Package, 10 L

Representative image only.

Reagents Potassium Hydroxide, 0.100N, Standardized, Cubitainer® Package, 10 L

Reagents | Mfr: C7285050-10F| Item: 6228801

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • N/10, In Methanol
  • 0.0995 to 0.1005 Normal
  • Colorless liquid
  • Density: 0.8
  • Boiling point: approximately 64.5 °C (148 °F)
  • Flash point: 11 °C (51.8 °F)
  • Solubility information: miscible
  • pH range: 13
  • 10 L Cubitainer® package

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Normality0.1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer® package
  • DOT InformationFLAMMABLE LIQUIDS, N.O.S. (METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1230
  • Shelf Life (days)395
  • Shelf Life (months)12
  • DescriptionPotassium Hydroxide, 0.100N, Standardized, Cubitainer® Package, 10 L
Documents & Certificates
Reviews
Q&A