Reagents Potassium Dichromate, ACS Reagent, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Potassium Dichromate, ACS Reagent, Natural Poly Container, 2.5 kg

Reagents | Mfr: C1282500-2.5A5| Item: 6228240

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Orange-red crystal
  • Density: 2.68
  • Solubility information: 151 g/L at 25 °C (77 °F)
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaK2Cr2O7
  • ClassSalts
  • ChemistryInorganics
  • SynonymsPotassium bichromate
  • FormSolid
  • Weight (g)2500
  • Formula Weight294.18
  • PackagingPoly container
  • DOT InformationTOXIC SOLIDS, OXIDIZING, N.O.S. (POTASSIUM DICHROMATE), 6.1 (5.1), P.G. I
  • Storage SymbolAmbient
  • CAS Number7778-50-9
  • UN NumberUN3086
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)500
  • Boiling Point F (° F)932
  • Melting Point C (° C)398
  • Melting Point F (° F)748.4
  • DescriptionPotassium Dichromate, ACS Reagent, Natural Poly Container, 2.5 kg
Documents & Certificates
Reviews
Q&A