Reagents Potassium Chromate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Representative image only.

Reagents Potassium Chromate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Reagents | Mfr: C1281000-125D2| Item: 6228235

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99%
  • Yellow crystal
  • Density: 2.73
  • Solubility information: 650 g/L at 25 °C (77 °F)
  • pH range: 8.6 to 9.8
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaK2CrO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • Formula Weight194.19
  • PackagingWide-mouth amber glass bottle
  • DOT InformationTOXIC, N.O.S. (POTASSIUM CHROMATE), 5.1 (6.1), P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3288
  • CAS Number7789-00-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)971
  • Melting Point F (° F)1779.8
  • DescriptionPotassium Chromate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g
Documents & Certificates
Reviews
Q&A