Reagents Potassium Chloride, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Potassium Chloride, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2280500-2.5A5| Item: 6228544

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.98
  • Solubility information: 347 g/L at 20 °C (68 °F)
  • pH range: 5.4 to 8.6
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaKCl
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)2500
  • Formula Weight74.55
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7447-40-7
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)1413
  • Boiling Point F (° F)2575.4
  • Melting Point C (° C)771
  • Melting Point F (° F)1419.8
  • DescriptionPotassium Chloride, Purified, Natural Poly Container, 2.5 kg
Documents & Certificates
Reviews
Q&A