Reagents Phthalic Anhydride, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Phthalic Anhydride, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2274000-2.5A5| Item: 6228540

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • White solid
  • Density: 1.53
  • Flash point: 152 °C (305.6 °F)
  • Solubility nformation: 6.4 g/L at 20 °C (68 °F)
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaC6H4(CO)2O
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • Weight (g)2500
  • Formula Weight148.12
  • Vapor Pressure (torr)0.000203
  • PackagingPoly container
  • DOT InformationPHTHALIC ANHYDRIDE, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number85-44-9
  • UN NumberUN2214
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)284
  • Boiling Point F (° F)543.2
  • Melting Point C (° C)131
  • Melting Point F (° F)267.8
  • DescriptionPhthalic Anhydride, Natural Poly Container, 2.5 kg
Documents & Certificates
Reviews
Q&A