Reagents Phosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 4 L

Representative image only.

Reagents Phosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 4 L

Reagents | Mfr: C5101950-4C| Item: 6228645

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 85%
  • Colorless liquid
  • Density: 1.69
  • Solubility information: miscible
  • pH range: <1
  • 4 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaH3PO4
  • ClassAcid
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight98
  • PackagingAmber glass bottle
  • DOT InformationPHOSPHORIC ACID SOLUTION, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number7664-38-2
  • UN NumberUN1805
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)21
  • Melting Point F (° F)69.8
  • DescriptionPhosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A