Reagents Phenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 100 mL

Representative image only.

Reagents Phenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 100 mL

Reagents | Mfr: C84003NE-100A| Item: 6228858

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • In Methanol
  • Colorless liquid
  • Density: 0.8
  • Flash point: 11 °C (51.8 °F)
  • Solubility information: miscible
  • pH range: 8.0 (colorless) to 10.0 (pink)
  • 100 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.25% w/v in MeOH
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)100
  • PackagingPoly bottle
  • DOT InformationFLAMMABLE LIQUIDS, N.O.S. (METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1993
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionPhenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 100 mL
Documents & Certificates
Reviews
Q&A