Reagents Phenolphthalein, ACS Reagent, Natural Poly Container, 3 kg

Representative image only.

Reagents Phenolphthalein, ACS Reagent, Natural Poly Container, 3 kg

Reagents | Mfr: C2270000-3KG| Item: 6228538

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White to pale yellow powder
  • Density: 1.3
  • Melting pont: 258 to 262 °C (496.4 to 503.6 °F)
  • Solubility information: 0.4 g/L at 20 °C (68 °F)
  • pH range: 8.0 (colorless) to 10.0 (pink)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC20H14O4
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)3000
  • Formula Weight318.32
  • PackagingNatural poly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number77-09-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPhenolphthalein, ACS Reagent, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A