Reagents Phenol (Liquefied), Purified, Amber Glass Bottle, 1 L

Representative image only.

Reagents Phenol (Liquefied), Purified, Amber Glass Bottle, 1 L

Reagents | Mfr: C2269500-1Y| Item: 6228536

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly pink liquid
  • Density: 1.07
  • Melting point: 40 to 42 °C (104 to 107.6 °F)
  • Flash point: 79 °C (174.2 °F)
  • Solubility information: miscible
  • pH range: 3.5 to 4.2
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaC6H5OH
  • ClassPhenol
  • ChemistryAnalytical Reagents
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight94.11
  • Vapor Pressure (torr)16.55
  • PackagingAmber glass bottle
  • DOT InformationPHENOL SOLUTIONS, 6.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number108-95-2
  • UN NumberUN2821
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)182
  • Boiling Point F (° F)359.6
  • DescriptionPhenol (Liquefied), Purified, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A