Reagents Petroleum Ether, ACS Reagent, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Petroleum Ether, ACS Reagent, Amber Glass Bottle, 500 mL

Reagents | Mfr: C1199000-500C| Item: 6228176

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.65
  • Boiling point: 35 to 60 °C (95 to 140 °F)
  • Flash point: -18 °C (-0.4)
  • Solubility information: insoluble
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC6-16H12-34
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsLigroin, Benzin, Petroleum naphtha, Petroleum spirits
  • Volume (mL)500
  • Formula Weight82.2
  • PackagingAmber glass bottle
  • DOT InformationPETROLEUM DISTILLATES, N.O.S., 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number8032-32-4
  • UN NumberUN1268
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionPetroleum Ether, ACS Reagent, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A