Reagents Oxalic Acid Dihydrate, Purified, Poly Pail, 12 kg

Representative image only.

Reagents Oxalic Acid Dihydrate, Purified, Poly Pail, 12 kg

Reagents | Mfr: C2110500-12KG| Item: 6228351

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless/white crystals
  • Density: 1.65
  • Melting point: 104 to 106 °C (219.2 to 222.8 °F)
  • Solubility information: 140 g/L at 25 °C (77 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaC2H6O6·2H2O
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • SynonymsOxalic acid 2-hydrate
  • Weight (g)12000
  • Formula Weight162.1
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6153-56-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionOxalic Acid Dihydrate, Purified, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A