Reagents Oleic Acid, Purified, Amber Glass Bottle, 1 L

Representative image only.

Reagents Oleic Acid, Purified, Amber Glass Bottle, 1 L

Reagents | Mfr: C2110000-1C| Item: 6228350

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly yellow liquid
  • Density: 0.9
  • Boiling point: 194 to 195 °C (381.2 to 383 °F)
  • Flash point: 180 °C (356 °F)
  • Refractive index: 1.4585 to 1.4605nD
  • Solubility information: insoluble
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaC18H34O2
  • ClassAcid
  • Synonymscis-9-Octadecenoic acid
  • FormLiquid
  • Volume (mL)1000
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number112-80-1
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)16
  • Melting Point F (° F)60.8
  • DescriptionOleic Acid, Purified, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A