Reagents o-Cresol with 0.1% Ethanol, Amber Glass Bottle, 1 L

Representative image only.

Reagents o-Cresol with 0.1% Ethanol, Amber Glass Bottle, 1 L

Reagents | Mfr: C2186350-1Y| Item: 6228483

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly yellow or pink
  • Density: 1.04
  • Melting point: 29 to 31 °C (84.2 to 87.8 °F)
  • Solubility information: 26 g/L at 25 °C (77 °F)
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaC7H8O
  • ClassAlcohol
  • ChemistrySolvents
  • Synonymsortho-Cresol
  • FormLiquid
  • Volume (mL)1000
  • PackagingAmber glass bottle
  • DOT InformationCRESOLS, LIQUID, 6.1 (8), P.G. II
  • Storage SymbolAmbient
  • CAS Number95-48-7
  • UN NumberUN2076
  • Shelf Life (days)570
  • Shelf Life (months)18
  • Boiling Point C (° C)191
  • Boiling Point F (° F)375.8
  • Descriptiono-Cresol with 0.1% Ethanol, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A