Reagents Nitrobenzene, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Nitrobenzene, Amber Glass Bottle, 500 mL

Reagents | Mfr: C2257500-500C| Item: 6228529

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Pale yellow liquid
  • Density: 1.2
  • Boiling point: 211 to 212 °C (411.8 to 413.6 °F)
  • Melting point: 5.6 to 5.8 °C (42.08 to 42.44 °F)
  • Flash point: 88 °C (190.4 °F)
  • Refractive index: 1.550 to 1.552nD
  • Solubility information: 2 g/L at 20 °C (68 °F)
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaC6H5NO2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)500
  • Formula Weight123.11
  • Vapor Pressure (torr)0.195
  • PackagingAmber glass bottle
  • DOT InformationNITROBENZENE, 6.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number98-95-3
  • UN NumberUN1662
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionNitrobenzene, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A