Reagents Nitric Acid, 70%, Trace Metal, ACS Reagent, Poly Bottle, 2.5 L

Representative image only.

Reagents Nitric Acid, 70%, Trace Metal, ACS Reagent, Poly Bottle, 2.5 L

Reagents | Mfr: C5101650-2.5A| Item: 6228642

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Meets ACS specifications
  • Assay 68.00-70.00%
  • Colorless to light yellow liquid
  • Density: 1.416
  • Solubility information: miscible
  • pH range: <1
  • 2.5 L poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeTrace Metals, ACS
  • Linear FormulaHNO3
  • ClassAcid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight63.0128
  • PackagingPoly bottle
  • DOT InformationNITRIC ACID, 8, P.G.II
  • Storage SymbolAmbient
  • CAS Number7697-37-2
  • UN NumberUN2031
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)121
  • Boiling Point F (° F)249.8
  • Melting Point C (° C)-32
  • Melting Point F (° F)-25.6
  • DescriptionNitric Acid, 70%, Trace Metal, ACS Reagent, Poly Bottle, 2.5 L
Documents & Certificates
Reviews
Q&A