Reagents n-Pentane, Amber Glass Bottle, 500 mL

Representative image only.

Reagents n-Pentane, Amber Glass Bottle, 500 mL

Reagents | Mfr: C2265600-500C| Item: 6228533

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless liquid
  • Density: 0.63
  • Boiling point: 35 to 36 °C (95 to 96.8 °F)
  • Flash point: -49 °C (-56.2 °F)
  • Solubility information: <1 mg/L at 20 °C (68 °F)
  • 500 mL amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaC5H12
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsPentane
  • Volume (mL)500
  • Formula Weight72.15
  • Vapor Pressure (torr)385.5317
  • PackagingAmber glass bottle
  • DOT InformationPENTANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number109-66-0
  • UN NumberUN1265
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)-130
  • Melting Point F (° F)-202
  • Refractive Index1.35
  • Descriptionn-Pentane, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A