Reagents N,N-Dimethylformamide, Purified, Drum, 200 L

Representative image only.

Reagents N,N-Dimethylformamide, Purified, Drum, 200 L

Reagents | Mfr: C2196200-200E| Item: 6228496

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless to slightly yellow liquid
  • Density: 0.94
  • Flash point: 58 °C (136.4 °F)
  • Refractive index: 1.429 to 1.432nD
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaHCON(CH3)2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDimethylformamide, DMF, Formyldimethylamine, Formamide, N,N-Dimethylformamide
  • Volume (mL)200000
  • Formula Weight73.09
  • Vapor Pressure (torr)0.0263
  • PackagingDrum
  • DOT InformationN,N-DIMETHYLFORMAMIDE, 3, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN2265
  • CAS Number68-12-2
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)153
  • Boiling Point F (° F)307.4
  • Melting Point C (° C)-61
  • Melting Point F (° F)-77.8
  • DescriptionN,N-Dimethylformamide, Purified, Drum, 200 L
Documents & Certificates
Reviews
Q&A