Reagents N,N-Dimethylacetamide, Drum, 15 gal.

Representative image only.

Reagents N,N-Dimethylacetamide, Drum, 15 gal.

Reagents | Mfr: C1196100-15E| Item: 6228171

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 0.94
  • Boiling point: 164.5 to 166 °C (328.1 to 330.8 °F)
  • Refractive index: 1.437 to 1.439nD
  • Solubility information: miscible
  • 15 gallon drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaCH3CN(CH3)2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDimethylacetamide, DMA
  • Formula Weight71.12
  • Volume (Gal)15
  • Vapor Pressure (torr)1.3201
  • PackagingDrum container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number127-19-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-20
  • Melting Point F (° F)-4
  • DescriptionN,N-Dimethylacetamide, Drum Container, 15 gal.
Documents & Certificates
Reviews
Q&A