Reagents n-Butyl Alcohol, Purified, Amber Glass Bottle, 4 L

Representative image only.

Reagents n-Butyl Alcohol, Purified, Amber Glass Bottle, 4 L

Reagents | Mfr: C2120000-4C| Item: 6228367

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.81
  • Refractive index: 1.398 to 1.400nD
  • Flash point: 37 °C (98.6 °F)
  • Solubility information: 70 g/L at 25 °C (77 °F)
  • 4 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaCH3(CH2)3OH
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1-Butanol
  • Volume (mL)4000
  • Formula Weight74.12
  • Vapor Pressure (torr)0.4935
  • PackagingAmber glass bottle
  • DOT InformationBUTANOLS, 3, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1120
  • CAS Number71-36-3
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)118
  • Boiling Point F (° F)244.4
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • Descriptionn-Butyl Alcohol, Purified, Amber Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A