Reagents n-Butyl Acetate, ASTM D4615, Amber Glass Bottle, 500 mL

Representative image only.

Reagents n-Butyl Acetate, ASTM D4615, Amber Glass Bottle, 500 mL

Reagents | Mfr: C2156300-500C| Item: 6228447

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.88
  • Boiling point: 124 to 126 °C (255.2 to 258.8 °F)
  • Flash point: 22 °C (71.6 °F)
  • Refractive index: 1.393 to 1.395nD
  • Solubility information: 7 g/L at 20 °C (68 °F)
  • Meets ASTM D4615
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeASTM
  • Linear FormulaC6H12O2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)500
  • Formula Weight116.16
  • Vapor Pressure (torr)9.7508
  • PackagingAmber glass bottle
  • DOT InformationBUTYL ACETATES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number123-86-4
  • UN NumberUN1123
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-78
  • Melting Point F (° F)-108.4
  • Descriptionn-Butyl Acetate, ASTM D4615, Amber Glass Bottle, 500 mL
Documents & Certificates
Reviews
Q&A