Reagents Murexide Indicator, Wide-Mouth Amber Glass Bottle, 5 g

Representative image only.

Reagents Murexide Indicator, Wide-Mouth Amber Glass Bottle, 5 g

Reagents | Mfr: C2251980-5C2| Item: 6228524

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Purple-red solid
  • Density: 1.9
  • Solubility information: slightly soluble
  • 5 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaC8H8N6O6
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Synonyms5,5′-Nitrilodibarbituric acid monoammonium salt, Ammonium purpurate
  • Weight (g)5
  • Formula Weight284.19
  • Vapor Pressure (torr)<1
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number3051-09-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)770
  • Melting Point F (° F)1418
  • DescriptionMurexide Indicator, Wide-Mouth Amber Glass Bottle, 5 g
Documents & Certificates
Reviews
Q&A