Reagents Methyl Red Indicator, 0.1% (w/v) in Ethanol, Natural Poly Bottle, 120 mL

Representative image only.

Reagents Methyl Red Indicator, 0.1% (w/v) in Ethanol, Natural Poly Bottle, 120 mL

Reagents | Mfr: C7118610-120A| Item: 6228771

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • In Ethanol
  • Red liquid
  • Density: Boiling point: approximately 77 °C (170.6 °F)
  • Melting point: approximately -90 °C (-130 °F)
  • Flash point: 14 °C (57.2 °F)
  • Solubility information: miscible
  • pH range: 4.2 (pink) to 6.2 (yellow)
  • 120 mL natural poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration0.1% (w/v) in ethanol
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)120
  • PackagingPoly bottle
  • DOT InformationETHANOL SOLUTION, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1170
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionMethyl Red Indicator, 0.1% (w/v) in Ethanol, Natural Poly Bottle, 120 mL
Documents & Certificates
Reviews
Q&A